GSK-2983559 98%

Supplier: Ambeed
Warning

Synonyms: 2-((4-(benzo[d]thiazol-5-ylamino)-6-(tert-butylsulphonyl)quinazolin-7-yl)oxy)ethyl dihydrogen phosphate, GSK2983559

A939659-10MG A939659-1MG A939659-5MG
77256-718EA 149.35 USD
77256-718 77256-720 77256-722
GSK-2983559 98%
GSK-2983559

Precaución: for Research Use Only

Formula: C₂₁H₂₃N₄O₇PS₂
MW: 538.53 g/mol
MDL Number: MFCD30489430
CAS Number: 1579965-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR