4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline 98%

Supplier: Ambeed
Warning

A939329-1G A939329-250MG A939329-5G
77294-310EA 33.99 USD
77294-310 77294-312 77294-314
4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline 98%
4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₄ClF₄N
MW: 249.5921 g/mol
MDL Number: MFCD01570536
CAS Number: 401567-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR