1-(5-Fluoro-2-methyl-3-nitrophenyl)ethanone 98%

Supplier: Ambeed
Warning

A938996-100MG A938996-250MG A938996-1G
77268-996EA 49.44 USD
77268-996 77268-998 77749-764
1-(5-Fluoro-2-methyl-3-nitrophenyl)ethanone 98%
1-(5-Fluoro-2-methyl-3-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈FNO₃
MW: 197.16 g/mol
MDL Number: MFCD20526343
CAS Number: 176548-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR