4-Bromo-2H-1,2,3-triazole 95%

Supplier: Ambeed
Warning

A937957-100MG A937957-250MG A937957-1G A937957-5G
77268-992EA 50 USD
77268-992 77268-994 77749-758 77749-759
4-Bromo-2H-1,2,3-triazole 95%
4-Bromo-2H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₂H₂BrN₃
MDL Number: MFCD28969562
CAS Number: 1988796-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR