dichlororuthenium,1,10-phenanthroline xhydrate 98%

Supplier: Ambeed
Warning

A936748-1G A936748-250MG A936748-5G A936748-100MG
77256-712EA 84.46 USD
77256-712 77256-714 77256-716 77747-700
dichlororuthenium,1,10-phenanthroline xhydrate 98%
dichlororuthenium,1,10-phenanthroline xhydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₀Cl₂N₂ORu
MW: 370.19 g/mol
MDL Number: MFCD00799695
CAS Number: 207802-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR