1-(6-Bromopyridin-2-yl)ethanamine 97%

Supplier: Ambeed
Danger

A936732-100MG A936732-10G A936732-5G A936732-1G
77256-706EA 99.91 USD
77256-706 77256-708 77256-710 77618-592
1-(6-Bromopyridin-2-yl)ethanamine 97%
1-(6-Bromopyridin-2-yl)ethanamine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MW: 201.06 g/mol
MDL Number: MFCD13188876
CAS Number: 914950-77-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR