2,3-Difluoro-4-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A936413-1G A936413-250MG
77256-702EA 16 USD
77256-702 77256-704
2,3-Difluoro-4-methoxybenzaldehyde 98%
2,3-Difluoro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD04115981
CAS Number: 256417-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR