Dde-Orn(Fmoc)-OH 98% (by HPLC)

Supplier: Ambeed
Warning

A935976-1G A935976-250MG A935976-5G
77244-840EA 117.42 USD
77244-840 77244-842 77244-844
Dde-Orn(Fmoc)-OH 98% (by HPLC)
Dde-Orn(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₃₀H₃₄N₂O₆
MW: 518.60 g/mol
MDL Number: MFCD06796885
CAS Number: 1423017-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR