4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-(methylsulphanyl)-6-(trifluoromethyl)pyrimidine

A935892-1G A935892-250MG A935892-5G
77294-298EA 21.63 USD
77294-298 77294-300 77294-302
4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine 97%
4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂S
MW: 228.617 g/mol
MDL Number: MFCD00269080
CAS Number: 16097-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR