(R)-4-(tert-butyl)-2-(2-(diphenylphosphino)phenyl)-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

A935663-100MG A935663-1G A935663-5G A935663-250MG
77256-698EA 47.38 USD
77256-698 77256-700 77595-514 77618-590
(R)-4-(tert-butyl)-2-(2-(diphenylphosphino)phenyl)-4,5-dihydrooxazole ≥98%, ee 99%
(R)-4-(tert-butyl)-2-(2-(diphenylphosphino)phenyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₅H₂₆NOP
MW: 387.45 g/mol
MDL Number: MFCD06799049
CAS Number: 164858-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR