Ethyl 2-hydroxy-5-(trifluoromethyl)benzoate 97%

Supplier: Ambeed
Warning

A935377-1G A935377-5G
77268-982EA 285.31 USD
77268-982 77268-984
Ethyl 2-hydroxy-5-(trifluoromethyl)benzoate 97%
Ethyl 2-hydroxy-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
MDL Number: MFCD18396569
CAS Number: 1261620-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR