Ethane-1,2-diylbis(phenylphosphinic acid) 95%

Supplier: Ambeed
Warning

A934650-100MG A934650-250MG A934650-50MG
77268-976EA 238.96 USD
77268-976 77268-978 77268-980
Ethane-1,2-diylbis(phenylphosphinic acid) 95%
Ethane-1,2-diylbis(phenylphosphinic acid)

Precaución: for Research Use Only

Formula: C₁₄H₁₆O₄P₂
MW: 310.22 g/mol
MDL Number: MFCD30607312
CAS Number: 1089-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR