2,6,10-Trimethyl-2,6,10-triazaundecane 98%

Supplier: Ambeed
Danger

Synonyms: N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine

A934476-5G A934476-25G
77256-690EA 5.15 USD
77256-690 77747-697
2,6,10-Trimethyl-2,6,10-triazaundecane 98%
2,6,10-Trimethyl-2,6,10-triazaundecane

Precaución: for Research Use Only

Formula: C₁₁H₂₇N₃
MW: 201.36 g/mol
MDL Number: MFCD00126936
CAS Number: 3855-32-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR