(4-(4-Methylthiazol-5-yl)phenyl)methanamine 95%

Supplier: Ambeed
Warning

A933581-5G A933581-1G A933581-250MG
77268-974EA 914.64 USD
77268-974 77569-658 77618-588
(4-(4-Methylthiazol-5-yl)phenyl)methanamine 95%
(4-(4-Methylthiazol-5-yl)phenyl)methanamine

Precaución: For research use only.

Formula: C₁₁H₁₂N₂S
MW: 204.29 g/mol
MDL Number: MFCD28965258
CAS Number: 1448189-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR