3-Methyl-2-butenal 97%

Supplier: Ambeed
Danger

Synonyms: 3,3-Dimethylacrolein, 3-Methylbut-2-enal, Senecialdehyde, 3-Methylcrotonaldehyde

A933415-10G A933415-1G A933415-25G A933415-5G
77281-978EA 23.69 USD
77281-978 77281-980 77281-982 77552-734
3-Methyl-2-butenal 97%
3-Methyl-2-butenal

Precaución: for Research Use Only

Formula: C₅H₈O
MW: 84.12 g/mol
MDL Number: MFCD00010291
CAS Number: 107-86-8
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR