(E)-2-Ethylhexyl 3-(4-methoxyphenyl)acrylate 98+%

Supplier: Ambeed
Warning

A932097-1000G A932097-25G A932097-500G A932097-100G
77294-292EA 246.17 USD
77294-292 77294-294 77294-296 77749-752
(E)-2-Ethylhexyl 3-(4-methoxyphenyl)acrylate 98+%
(E)-2-Ethylhexyl 3-(4-methoxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₈H₂₆O₃
MW: 290.39 g/mol
MDL Number: MFCD00072582
CAS Number: 83834-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR