1-Hexynylbenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-1-hexyne, 1-Butyl-2-phenylacetylene

A929993-1G A929993-250MG A929993-25G A929993-5G
77244-826EA 24.72 USD
77244-826 77244-828 77244-830 77244-832
1-Hexynylbenzene 98%
1-Hexynylbenzene

Precaución: for Research Use Only

Formula: C₆H₅CC(CH₂)₃CH₃
MW: 158.24 g/mol
MDL Number: MFCD00039969
CAS Number: 1129-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR