(3-Chloropropyl)triethoxysilane 97%

Supplier: Ambeed
Danger

Synonyms: 3-Chloropropyltriethoxysilane, Triethoxy(3-chloropropyl)silane, 3-(Triethoxysilyl)propylchloride

A929769-25G A929769-100G
77281-976EA 21 USD
77281-976 77749-751
(3-Chloropropyl)triethoxysilane 97%
(3-Chloropropyl)triethoxysilane

Precaución: for Research Use Only

Formula: ClH₂CCH₂CH₂Si(OCH₂CH₃)₃
MDL Number: MFCD00018985
CAS Number: 5089-70-3
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR