3-(2,3-Dimethoxyphenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2,3-Dimethoxyphenyl)propanoic acid

A929509-250MG A929509-25G
77281-974EA 6.18 USD
77281-974 77749-750
3-(2,3-Dimethoxyphenyl)propionic acid 98%
3-(2,3-Dimethoxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
MDL Number: MFCD00029821
CAS Number: 10538-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR