2-Hexyldecanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Isopalmitic Acid, Pentadecane-7-carboxylic Acid

A929174-25G A929174-500G A929174-5G A929174-100G
77268-956EA 25.75 USD
77268-956 77268-958 77268-960 77552-724
2-Hexyldecanoic acid 98%
2-Hexyldecanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
MDL Number: MFCD00059651
CAS Number: 25354-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR