2-Fluoro-9H-carbazole 99%

Supplier: Ambeed
Warning

A928793-1G A928793-250MG A928793-5G
77268-950EA 43 USD
77268-950 77268-952 77268-954
2-Fluoro-9H-carbazole 99%

Precaución: for Research Use Only

Formula: C₁₂H₈FN
MDL Number: MFCD09033508
CAS Number: 391-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR