(2-Carbamoylphenoxy)acetic acid 95%

Supplier: Ambeed
Warning

A928757-1G A928757-25G A928757-5G
77281-968EA 15 USD
77281-968 77281-970 77281-972
(2-Carbamoylphenoxy)acetic acid 95%
(2-Carbamoylphenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD00025472
CAS Number: 25395-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR