2-(3-Chloro-4-methoxyphenyl)acetonitrile 97%

Supplier: Ambeed
Warning

A928685-250MG A928685-5G A928685-1G
77281-964EA 44 USD
77281-964 77281-966 77618-582
2-(3-Chloro-4-methoxyphenyl)acetonitrile 97%
2-(3-Chloro-4-methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MDL Number: MFCD00068623
CAS Number: 7569-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR