N'-(Pyridin-4-ylmethylene)isonicotinohydrazide 97%

Supplier: Ambeed
Warning

A928314-100MG A928314-1G A928314-250MG
77256-648EA 30.9 USD
77256-648 77256-650 77256-652
N'-(Pyridin-4-ylmethylene)isonicotinohydrazide 97%
N'-(Pyridin-4-ylmethylene)isonicotinohydrazide

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₄O
MW: 226.23 g/mol
CAS Number: 13025-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR