(3-Phenoxyphenyl)methanamine 95%

Supplier: Ambeed
Warning

A926419-1G A926419-250MG
77294-276EA 238.96 USD
77294-276 77294-278
(3-Phenoxyphenyl)methanamine 95%
(3-Phenoxyphenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
MDL Number: MFCD01631862
CAS Number: 50742-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR