2,2-Bis(4-methylphenyl)hexafluoropropane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4'-(Hexafluoroisopropylidene)ditoluene, 4,4'-(Perfluoropropane-2,2-diyl)bis(methylbenzene)
A925916-1G
A925916-25G
77294-272EA
6.18
USD
77294-272
77294-274
2,2-Bis(4-methylphenyl)hexafluoropropane 98%
2,2-Bis(4-methylphenyl)hexafluoropropane
Precaución: for Research Use Only
Formula:
C₁₇H₁₄F₆ MW: 332.29 g/mol |
MDL Number:
MFCD00042597 CAS Number: 1095-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...