4-Decanol 97%

Supplier: Ambeed
Danger

Synonyms: Decan-4-ol

A925556-1G A925556-5G
77294-268EA 8.24 USD
77294-268 77294-270
4-Decanol 97%
4-Decanol

Precaución: for Research Use Only

Formula: C₁₀H₂₂O
MW: 158.28 g/mol
MDL Number: MFCD00039627
CAS Number: 2051-31-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR