4'-Aminobiphenyl-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Amino-[1,1'-biphenyl]-4-ol

A924846-100MG A924846-250MG A924846-5G A924846-1G
77294-262EA 33.99 USD
77294-262 77294-264 77294-266 77749-747
4'-Aminobiphenyl-4-ol 98%
4'-Aminobiphenyl-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
MDL Number: MFCD00463439
CAS Number: 1204-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR