4-Bromo-2-ethylaniline 98%

Supplier: Ambeed
Danger

A924432-10G A924432-1G A924432-25G A924432-5G
77294-254EA 28.84 USD
77294-254 77294-256 77294-258 77294-260
4-Bromo-2-ethylaniline 98%
4-Bromo-2-ethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD00041434
CAS Number: 45762-41-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR