AZD-2098 98+%

Supplier: Ambeed
Warning

A924249-10MG A924249-1MG A924249-5MG
77281-952EA 33 USD
77281-952 77281-954 77281-956
AZD-2098 98+%
AZD-2098

Precaución: for Research Use Only

Formula: C₁₁H₉Cl₂N₃O₃S
MDL Number: MFCD31382158
CAS Number: 566203-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR