N1,N1,2-Trimethylpropane-1,2-diamine dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
A924221-1G
A924221-250MG
A924221-5G
77268-928EA
65.92
USD
77268-928
77268-930
77569-646
N1,N1,2-Trimethylpropane-1,2-diamine dihydrochloride 97%
N1,N1,2-Trimethylpropane-1,2-diamine dihydrochloride
Precaución: For research use only.
Formula:
C₆H₁₈Cl₂N₂ MW: 189.12 g/mol |
MDL Number:
MFCD22631440 CAS Number: 75975-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...