2,4-Dibromobenzoic acid 97%

Supplier: Ambeed
Warning

A924129-100G A924129-10G A924129-1G A924129-25G
77256-632EA 315.18 USD
77256-632 77256-634 77256-636 77256-638
2,4-Dibromobenzoic acid 97%
2,4-Dibromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
MDL Number: MFCD00234253
CAS Number: 611-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR