trans-3-Methoxy-β-nitrostyrene 97%

Supplier: Ambeed
Warning

A922718-10G A922718-1G A922718-250MG
77244-800EA 107.12 USD
77244-800 77244-802 77244-804
trans-3-Methoxy-β-nitrostyrene 97%
trans-3-Methoxy-β-nitrostyrene

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.18 g/mol
MDL Number: MFCD00024822
CAS Number: 3179-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR