(S)-β-Phenylalaninol 98%

Supplier: Ambeed
Danger

Synonyms: (S)-3-Amino-3-phenyl-1-propanol, (S)-3-Amino-3-phenylpropan-1-ol

A922036-100G A922036-100MG A922036-10G A922036-1G A922036-25G A922036-5G A922036-250MG
77256-620EA 655.08 USD
77256-620 77256-622 77256-624 77256-626 77256-628 77256-630 77747-683
(S)-β-Phenylalaninol 98%
(S)-β-Phenylalaninol

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.2056 g/mol
MDL Number: MFCD01311768
CAS Number: 82769-76-4
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR