1-Phenacylpyridinium bromide 98%

Supplier: Ambeed
Warning

A921559-100G A921559-1G A921559-25G A921559-5G
77244-792EA 115 USD
77244-792 77244-794 77244-796 77244-798
1-Phenacylpyridinium bromide 98%
1-Phenacylpyridinium bromide

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrNO
MDL Number: MFCD00031995
CAS Number: 16883-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR