4-Allylpyrocatechol 98%

Supplier: Ambeed
Warning

A921166-100MG A921166-1G A921166-250MG
77281-946EA 99 USD
77281-946 77281-948 77281-950
4-Allylpyrocatechol 98%
4-Allylpyrocatechol

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD01548379
CAS Number: 1126-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR