6-Methoxy-2-naphthaleneboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-2-naphthylboronic acid, 6-Methoxy-2-naphthylboronic Acid

A921117-10G A921117-1G A921117-25G A921117-5G
77281-938EA 41.2 USD
77281-938 77281-940 77281-942 77281-944
6-Methoxy-2-naphthaleneboronic acid 96%
6-Methoxy-2-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BO₃
MW: 202.02 g/mol
MDL Number: MFCD03093087
CAS Number: 156641-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR