2-Ethoxynaphthalene 99%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-naphthyl ether

A921010-100G A921010-25G
77268-906EA 12.36 USD
77268-906 77268-908
2-Ethoxynaphthalene 99%
2-Ethoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MW: 172.23 g/mol
MDL Number: MFCD00016808
CAS Number: 93-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR