4-(1-Methyl-1H-pyrazol-3-yl)phenylamine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(1-Methyl-1H-pyrazol-3-yl)aniline

A919624-100MG A919624-250MG A919624-1G
77294-236EA 51.5 USD
77294-236 77294-238 77595-482
4-(1-Methyl-1H-pyrazol-3-yl)phenylamine 97%
4-(1-Methyl-1H-pyrazol-3-yl)phenylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃
MW: 173.22 g/mol
MDL Number: MFCD03407387
CAS Number: 916766-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR