Quinoline-2,7-diamine 97%

Supplier: Ambeed
Warning

A917188-100MG A917188-1G A917188-250MG
77244-786EA 88 USD
77244-786 77244-788 77244-790
Quinoline-2,7-diamine 97%
Quinoline-2,7-diamine

Precaución: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD18802697
CAS Number: 114058-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR