5-Methoxy-m-cresol 98%

Supplier: Ambeed
Warning

A916934-1G A916934-250MG A916934-25G
77281-918EA 40 USD
77281-918 77281-920 77281-922
5-Methoxy-m-cresol 98%
5-Methoxy-m-cresol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MDL Number: MFCD00059261
CAS Number: 3209-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR