1-Phenyl-3,4-dihydroisoquinoline 95%

Supplier: Ambeed
Warning

A915943-1G A915943-250MG A915943-5G
77244-778EA 45 USD
77244-778 77244-780 77244-782
1-Phenyl-3,4-dihydroisoquinoline 95%
1-Phenyl-3,4-dihydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₅H₁₃N
MDL Number: MFCD01817309
CAS Number: 52250-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR