3-Methylbenzo[d]oxazole-2(3H)-thione 95%

Supplier: Ambeed
Warning

A915382-1G A915382-250MG A915382-5G
77281-912EA 62.83 USD
77281-912 77281-914 77281-916
3-Methylbenzo[d]oxazole-2(3H)-thione 95%
3-Methylbenzo[d]oxazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₈H₇NOS
MW: 165.21 g/mol
MDL Number: MFCD00459943
CAS Number: 13673-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR