4-(3-Chloro-2-pyridinyl)morpholine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-2-morpholinopyridine
A915066-1G
A915066-5G
77281-908EA
37.08
USD
77281-908
77281-910
4-(3-Chloro-2-pyridinyl)morpholine 98%
4-(3-Chloro-2-pyridinyl)morpholine
Precaución: for Research Use Only
Formula:
C₉H₁₁ClN₂O MW: 198.65 g/mol |
MDL Number:
MFCD00498941 CAS Number: 54231-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...