Posaconazole inter-4 95%

Supplier: Ambeed
Warning

Synonyms: ((2S,3S)-2-(Benzyloxy)pentan-3-yl)hydrazine (2S,3S)-2,3-bis(benzoyloxy)succinate (X:1)

A913769-250MG A913769-5G
77268-886EA 7 USD
77268-886 77749-729
Posaconazole inter-4 95%
Posaconazole inter-4

Precaución: for Research Use Only

Formula: C₃₀H₃₄N₂O₉
MDL Number: MFCD22124447
CAS Number: 183871-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR