N4-(Benzo[d][1,3]dioxol-5-ylmethyl)-6-(3-methoxyphenyl)pyrimidine-2,4-diamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: BML-284 HCl

A913715-10MG A913715-1MG A913715-25MG A913715-50MG A913715-5MG
77268-876EA 80.34 USD
77268-876 77268-878 77268-880 77268-882 77268-884
N4-(Benzo[d][1,3]dioxol-5-ylmethyl)-6-(3-methoxyphenyl)pyrimidine-2,4-diamine hydrochloride 98%
N4-(Benzo[d][1,3]dioxol-5-ylmethyl)-6-(3-methoxyphenyl)pyrimidine-2,4-diamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₁₉ClN₄O₃
MW: 386.83 g/mol
MDL Number: MFCD26142942
CAS Number: 2095432-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR