3-(N-Ethylaminocarbonyl)-5-nitrobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Ethylcarbamoyl)-5-nitrophenylboronic acid

A913308-1G A913308-250MG A913308-5G
77256-582EA 76.22 USD
77256-582 77256-584 77256-586
3-(N-Ethylaminocarbonyl)-5-nitrobenzeneboronic acid 98%
3-(N-Ethylaminocarbonyl)-5-nitrobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BN₂O₅
MW: 238.005 g/mol
MDL Number: MFCD07783872
CAS Number: 871332-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR