(S,S)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride 98%
Supplier: Ambeed
|
Warning
|
A913279-100MG
A913279-1G
A913279-5G
77256-576EA
21
USD
77256-576
77256-578
77256-580
(S,S)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride 98%
(S,S)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride
Precaución: for Research Use Only
Formula:
C₃₆H₅₂AlClN₂O₂ |
MDL Number:
MFCD02683455 CAS Number: 307926-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...