2,6-Dimethyl-p-benzoquinone 98%

Supplier: Ambeed
Warning

A913066-1G A913066-25G A913066-5G
77256-570EA 12 USD
77256-570 77256-572 77256-574
2,6-Dimethyl-p-benzoquinone 98%
2,6-Dimethyl-p-benzoquinone

Precaución: for Research Use Only

Formula: C₈H₈O₂
MDL Number: MFCD00001605
CAS Number: 527-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR