3-tert-Butyl-4-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A913060-250MG A913060-1G
77281-906EA 50 USD
77281-906 77618-544
3-tert-Butyl-4-hydroxybenzaldehyde 95%
3-tert-Butyl-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD11848619
CAS Number: 65678-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR